Please login to your online account to display your discounted pricing

Salermide, Tocris™

SIRT1 and SIRT2 inhibitor

$142.08 - $598.33

Chemical Identifiers

Chemical Name or Material Salermide
Assay Percent Range >98%
Molecular Formula C26H22N2O2
Formula Weight 394.47
Synonym N-[3-[[(2-Hydroxy-1-naphthalenyl)methylene]amino]phenyl]-α-methylbenzeneacetamide
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

412710

 
r & d systems
RDS412710
10mg Each for $142.08

412750

 
r & d systems
RDS412750
50mg Each for $598.33
Description & Specifications

Specifications

Formula Weight 394.47
Quantity 10mg
Molecular Formula C26H22N2O2
Chemical Name or Material Salermide
Assay Percent Range >98%
CAS 1105698-15-4
Synonym N-[3-[[(2-Hydroxy-1-naphthalenyl)methylene]amino]phenyl]-α-methylbenzeneacetamide
Recommended Storage Store at +4°C
Safety and Handling

Store at +4°C