missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Salcomine 95.0+%, TCI America™

Manufacturer:  TCI America S0318100G

 View more versions of this product

Catalog No. S0318100G



Specifications

Specifications

Salcomine [=N,N′-Bis(salicylidene)ethylenediiminocobalt(II)]
100g
C16H16CoN2O2
N,N′-Bis(salicylidene)ethylenediiminocobalt(II), N,N′-Disalicylalethylenediamine Cobalt(II), N,N′-Ethylenebis(salicylideneiminato)cobalt(II)
C1=CC(=CNCCNC=C2C=CC=CC2=O)C(=O)C=C1.[Co]
327.249
325.23
≥95.0% (T)
Undesignated
14167-18-1
MFCD00000009
FIONSUNUNBIGCA-QVGGGXJLSA-N
cobalt;(6Z)-6-[[2-[[(E)-(6-oxocyclohexa-2,4-dien-1-ylidene)methyl]amino]ethylamino]methylidene]cyclohexa-2,4-dien-1-one
57448789
Crystal-Powder at 20°C

Chemical Identifiers

14167-18-1
327.249
FIONSUNUNBIGCA-QVGGGXJLSA-N
57448789
C1=CC(=CNCCNC=C2C=CC=CC2=O)C(=O)C=C1.[Co]
C16H16CoN2O2
MFCD00000009
N,N′-Bis(salicylidene)ethylenediiminocobalt(II), N,N′-Disalicylalethylenediamine Cobalt(II), N,N′-Ethylenebis(salicylideneiminato)cobalt(II)
cobalt;(6Z)-6-[[2-[[(E)-(6-oxocyclohexa-2,4-dien-1-ylidene)methyl]amino]ethylamino]methylidene]cyclohexa-2,4-dien-1-one
Safety and Handling

Safety and Handling

RTECSNumber : GG0590000

TSCA : Yes

Documents