Please login to your online account to display your discounted pricing

Salbutamol, 98%

A beta-adrenergic agonist

Specifications

Assay 0.99
CAS 18559-94-9
Formula Weight 239.31g/mol
Synonym Albuterol
Format Powder
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAJ63741MC

 
alfa aesar™
J63741MC
CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAJ63741MD

 
alfa aesar™
J63741MD
CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Assay 0.99
CAS 18559-94-9
Formula Weight 239.31g/mol
Synonym Albuterol
Format Powder
For Use With (Application) A beta-adrenergic agonist
Description White to Off-white
Melting Point 157° to 160°C