Please login to your online account to display your discounted pricing

Tocris™ Sal 003

Cell-permeable inhibitor of eIF2alpha dephosphorylation

$208.25 - $867.95

Specifications

Product Type Sal 003
Inhibitors Sal 003
Molecular Weight 463.21
Purity >99
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Quantity Price Quantity    

365710R

 
tocris™
RDS365710
10mg Each for $208.25

365750R

 
tocris™
RDS365750
50mg Each for $867.95
Description & Specifications

Specifications

Product Type Sal 003
Inhibitors Sal 003
Molecular Weight 463.21
Purity >99

Cell-permeable inhibitor of cellular phosphatase complexes that dephosphorylate eukaryotic translation initiation factor 2 subunit α (eIF2α). Analog of salubrinal (Cat. No. 2347) with improved aqueous solubility. Shown to prevent the induction of hippocampal long-term potentiation (LTP) and memory formation (LTM) in mice.

  • Chemical Name: 3--Phenyl-N-(2,2,2-trichloro-1-((((4-chlorophenyl)amino)carbonothioyl)amino)ethyl)acrylamide
  • Formula: C18H15Cl4N3OS
  • CAS Number: 1164470-53-4
  • Solubility: Soluble to 100mM in DMSO
  • Purity: >99