Saint-Gobain™ Tygon™ Sanitary Silicone Tubing Formula 3350
Translucent tubing ideal for biotechnological, pharmaceutical, and peristaltic applications
Manufacturer:  Saint-Gobain™ ABW00027
 View more versions of this product
Catalog No. 02-587-1V
Description
Description
Translucent tubing ideal for biotechnological, pharmaceutical, and peristaltic applications
Order Info
Packaged in heat-sealed, double-wrapped polyethylene packs of 50 ft. (15.2cm)
Compliance
Complies with requirements of USP XXII Class VI criteria. Meets 3A Sanitary Standards, FDA 21 CFR Part 177.2600 criteria and NSF 51 Standard.
Specifications
Specifications
Complies with requirements of USP XXII Class VI criteria. Meets 3A Sanitary Standards, FDA 21 CFR Part 177.2600 criteria and NSF 51 Standard.
3/8 in.
Tygon
12.7mm
0.0625 in.
Yes
9.5mm
1/2 in.
50 ft.
1.6mm
Documents
Accessories