Saint-Gobain™ Tygon S3™ E-LFL Non-DEHP Tubing
$273.82 - $319.85
Specifications
Length (English) 25 ft.
Length (Metric) 7.62m
View More Specs
Products ${productFamilyLength}
Catalog Number Mfr. No. Diameter (Metric) Inner Diameter (Metric) Outer Pressure Price Quantity  
Catalog Number Mfr. No. Diameter (Metric) Inner Diameter (Metric) Outer Pressure Price Quantity  
14-171-101 Saint-Gobain™
AVX42022
7.9mm 11.1mm 16 psi Pack of 25 Each for $319.85
14-171-100 Saint-Gobain™
AVX42017
6.4mm 9.5mm 20 psi Pack of 25 Each for $273.82
Specifications Description & Specifications
Specifications
25 ft.
7.62m