Saint-Gobain™ Tygon S3™ E-3603 Flexible Tubing 500 ft.
$236.88 - $1,159.21
Specifications
Material TygonS3
Length (English) 500 ft.
View More Specs
Products ${productFamilyLength}
Catalog Number Mfr. No. Diameter (English) Inner Diameter (English) Outer Thickness (English) Wall Price Quantity  
Catalog Number Mfr. No. Diameter (English) Inner Diameter (English) Outer Thickness (English) Wall Price Quantity  
14-171-269 Saint-Gobain™
ACF04006
1/8 in. 3/16 in. 1/32 in. Pack of 500 Each for $362.55
14-171-268 Saint-Gobain™
ACF04002
1/16 in. 1/8 in. 1/32 in. Pack of 500 Each for $236.88
14-171-270 Saint-Gobain™
ACF04007
1/8 in. 1/4 in. 1/16 in. Pack of 500 Each for $833.42
14-171-271 Saint-Gobain™
ACF04012
3/16 in. 5/16 in. 1/16 in. Pack of 500 Each for $1,159.21
Specifications Description & Specifications
Specifications
TygonS3
500 ft.