Saint-Gobain™ Tygon S3™ Clear Tubing, Formula B-44-4X, Non-DEHP
Specially designed to ensure bacterial-free fluid path for food processing applications
$57.18 - $669.12
Specifications
Color Clear
Maximum Pressure 9.5MPa
View More Specs
Products 8
Catalog Number Mfr. No. Diameter (English) Inner Diameter (English) Outer Price Quantity  
Catalog Number Mfr. No. Diameter (English) Inner Diameter (English) Outer Price Quantity  
14-171-145 View Documents Saint-Gobain™
AAA00010
5/32 in. 9/32 in. Pack of 50 Each for $117.09
14-171-148 View Documents Saint-Gobain™
AAA00017
1/4 in. 3/8 in. Pack of 50 Each for $167.30
14-171-147 View Documents Saint-Gobain™
AAA00012
3/16 in. 5/16 in. Pack of 50 Each for $146.52
14-171-146 View Documents Saint-Gobain™
AAA00011
3/16 in. 1/4 in. Pack of 50 Each for $57.18
14-171-149 View Documents Saint-Gobain™
AAA00018NONDEHP
1/4 in. 7/16 in. Pack for $236.64
14-171-156 View Documents Saint-Gobain™
AAA00029
3/8 in. 5/8 in. Pack of 50 Each for $448.97
14-171-161 View Documents Saint-Gobain™
AAA00039
1/2 in. 13/16 in. Pack of 50 Each for $649.39
14-171-164 View Documents Saint-Gobain™
AAA00053
3/4 in. 1 in. Pack of 50 Each for $669.12
Description
Description
  • Made with phthalate-free, non-DEHP flexible plastic
  • Smooth and non-porous texture prevent from particle entrapment to avoid bacterial growth
  • Good resistance to harsh alkaline cleaners and remain unaffected by common sanitizers
  • Compatible with food that contain high oil
  • Lightweight and easy to handle with minimum requirements of couplings and fittings
  • Available in large bore sizes up to 6 in. inside diameter
  • Provides clear visual inspection and flow control
  • Reinforced tubing available for elevated pressure
Aseptic filling, Condiment dispensing, Dairy processing, Vitamin and flavor concentrate system, Soft-serve dispensing
Compliance
FDA, NSF, 3-A, Japan Food Sanitation Law no. 370/1959, REACH, 1935/2004/EC and 10/2011/EU
Specifications
Specifications
Clear
9.5MPa