Please login to your online account to display your discounted pricing

Saint-Gobain™ Tygon S3™ B-44-4X I.B. Tubing

$434.61 - $3,779.85

Specifications

Color Clear
Material Phthalate-free plastic
For Use With (Application) Food, dairy, cosmetics
View More Specs
Products
Catalog Number Mfr. No. Diameter (English) Inner Diameter (English) Outer Length (English) Pressure Price Quantity    

14-171-107

 
saint-gobain™
ADA00038
1/2 in. 3/4 in. 50 in. 200 psi Pack of 50 Each for $573.36

14-171-106

 
saint-gobain™
ADA00029
3/8 in. 5/8 in. 50 in. 170 psi Pack of 50 Each for $434.61

14-171-108

 
saint-gobain™
ADA00046
5/8 in. 7/8 in. 50 in. 165 psi Pack of 50 Each for $729.09

14-171-109

 
saint-gobain™
ADA00053
3/4 in. 1 in. 50 in. 145 psi Pack of 50 Each for $800.30

14-171-112

 
saint-gobain™
ADA05074
1-1/2 in. 2 in. 20 in. 75 psi Pack of 20 Each for $1,253.73

14-171-115

 
saint-gobain™
ADAUN053
3/4 in. 1 in. 10/10 ft. Not Applicable Pack of 100 Each for $1,858.58

14-171-114

 
saint-gobain™
ADAUN029
3/8 in. 5/8 in. 10/10 ft. Not Applicable Pack of 100 Each for $1,012.88

14-171-113

 
saint-gobain™
ADA05086
3 in. 3-3/4 in. 20 in. 40 psi Pack of 20 Each for $3,779.85
Description & Specifications

Specifications

Color Clear
Material Phthalate-free plastic
For Use With (Application) Food, dairy, cosmetics