missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Saint-Gobain™ Tygon™ R-3603 Clear Laboratory Tubing - 100 ft.

Crystal clear and flexible tubing with very high chemical resistance

$704.24 - $704.24



Products ${productFamilyLength}
Catalog Number Mfr. No. Price Quantity  
Catalog Number Mfr. No. Price Quantity  
14-168-44 Saint-Gobain™
ACF02019
Pack for $704.24
Specifications

Description

  • Handles virtually all inorganic chemicals found in the lab
  • Nonoxidizing and noncontaminating
  • Long lasting and crack-resistant
  • Remains flexible at -43°C (-45°F)
  • Resists dry heat to 70°C (158°F)
  • Foot markings at 3 ft. intervals
  • Durometer hardness: Shore A, 55

Compliance

Meets FDA CFR Part 175.300 criteria