Saint-Gobain™ Tygon™ R-3400 UV Resistant Tubing
For virtually any permanent or temporary chemical transfer application
$204.24 - $204.24
Specifications
Material Tygon
View More Specs
Products ${productFamilyLength}
Catalog Number Mfr. No. Diameter (English) Inner Diameter (English) Outer Thickness Thickness (English) Wall Price Quantity  
Catalog Number Mfr. No. Diameter (English) Inner Diameter (English) Outer Thickness Thickness (English) Wall Price Quantity  
14-169-13J Saint-Gobain™
AAE00017
0.25 in. 0.38 in. 1.6mm (0.06 in.) 0.63 in. Pack of 50 Each for $204.24
Specifications
Description
  • Protects light-sensitive fluids
  • Wide range of chemical resistance
  • Resistant to ozone
  • Resistant to burn
  • Low compression set resists permanent deformation
  • Black
  • Available in clear formulation for fluid flow monitoring
  • Acid and caustic transfer
  • Corrosive diffusible gases
  • Fertilizer and pesticide dispensing
  • Electrical insulation lines
  • Ink and adhesive dispensing
  • General maintenance
Description & Specifications
Specifications
Tygon