missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Saint-Gobain Tygon™ ND 100-80 Tubing

Specifically formulated in a higher durameter that enables high resistance to kinking and collapse

$406.03 - $406.03

Specifications

Color Clear
For Use With (Application) Medical grade
View More Specs

Products 1
Catalog Number Mfr. No. Diameter (English) Inner Diameter (English) Outer Thickness (English) Price Quantity  
Catalog Number Mfr. No. Diameter (English) Inner Diameter (English) Outer Thickness (English) Price Quantity  
14-171-284 View Documents Saint-Gobain
AAD04103
0.02 in. 0.06 in. 0.02 in. Pack of 500 Each for $406.03
 
Description

Description

Specifically formulated in a higher durameter that enables high resistance to kinking and collapse

  • Designed for medical products where non-DEHP plastic material is required
  • Compatible with blood contact medical devices
  • Clear and glass-smooth inner bore for better visibility
  • Limited flexibility for suction applications
  • Available in five very small sizes for various surgical applications
  • Rigid enough for handling as well as soft enough to reduce puncturing
  • Sterilization methods: autoclavable, gas and radiation

Catheters, IV or intra-artrial infusion and other surgical applications

Compliance

UPS class VI approved and REACH Compliant.

Specifications

Specifications

Clear
Medical grade