Saint-Gobain Tygon™ ND 100-65 Medical Tubings
For blood contact medical devices where non-DEHP plasticized materials are required
$41.33 - $745.27
Specifications
Color Clear
Material Non-DEHP
Overall Length (English) 50 ft.
Overall Length (Metric) 15.2m
View More Specs
Products 12
Catalog Number Mfr. No. Diameter (English) Inner Diameter (English) Outer Thickness (English) Price Quantity  
Catalog Number Mfr. No. Diameter (English) Inner Diameter (English) Outer Thickness (English) Price Quantity  
14-171-295 View Documents Saint-Gobain
ADF00004
3/32 in. 5/32 in. 1/32 in. Pack of 50 Each for $41.33
14-171-298 View Documents Saint-Gobain
ADF00007
1/8 in. 1/4 in. 1/16 in. Pack of 50 Each for $122.79
14-171-302 View Documents Saint-Gobain
ADF00012
3/16 in. 5/16 in. 1/16 in. Pack of 50 Each for $160.70
14-171-306 View Documents Saint-Gobain
ADF00017
1/4 in. 3/8 in. 1/16 in. Pack of 50 Each for $216.67
14-171-307 View Documents Saint-Gobain
ADF00018
1/4 in. 7/16 in. 3/32 in. Pack of 50 Each for $331.21
14-171-309 View Documents Saint-Gobain
ADF00022
5/16 in. 7/16 in. 1/16 in. Pack of 50 Each for $235.15
14-171-308 View Documents Saint-Gobain
ADF00019
1/4 in. 1/2 in. 1/8 in. Pack of 50 Each for $396.36
14-171-312 View Documents Saint-Gobain
ADF00027
3/8 in. 1/2 in. 1/16 in. Pack of 50 Each for $273.48
14-171-313 View Documents Saint-Gobain
ADF00028
3/8 in. 9/16 in. 3/32 in. Pack of 50 Each for $376.70
14-171-317 View Documents Saint-Gobain
ADF00037
1/2 in. 11/16 in. 3/32 in. Pack of 50 Each for $492.03
14-171-316 View Documents Saint-Gobain
ADF00036
1/2 in. 5/8 in. 1/16 in. Pack of 50 Each for $332.97
14-171-318 View Documents Saint-Gobain
ADF00038
1/2 in. 3/4 in. 1/8 in. Pack of 50 Each for $745.27
Description
Description
  • Crystal clear for easy visualization of fluid flow
  • For contact with bodily fluids
  • Non-wetting surface permits complete drainage
  • Non-hemolytic and non-pyrogenic
  • Withstands EtO, gas and gamma sterilization
  • Minimally invasive devices
  • Blood and IV solutions
  • Dialysis equipment
  • Wound drainage
  • Inhalation equipment
  • Chemotherapy drug delivery
  • Kidney dialysis pumps
  • Peristaltic pumps
  • Catheters
Compliance
Meets USP Class VI criteria, USP Class VI, ISO 10993, European Pharmacopoeia 3.1.1.2 and European REACH requirements.
Specifications
Specifications
Clear
Non-DEHP
50 ft.
15.2m