Please login to your online account to display your discounted pricing
Saint-Gobain™ Tygon™ Microbore Tubing, Formula S-54-HL
Use as catheters for intravenous and arterial infusion
$399.58 - $601.33
Products
Catalog Number Mfr. No. Price Quantity    
14-170-15A
  Accessories
saint gobain perform plas
AAD04091
Pack for $514.55
14-170-15C
  Accessories
saint gobain perform plas
AAD04119
Pack for $399.58
14-170-15D
  Accessories
saint gobain perform plas
AAD04127
Pack for $571.33
14-170-15E
  Accessories
saint gobain perform plas
AAD04133
Pack for $601.33
Description & Specifications
  • Nontoxic, nonpyrogenic, and blood compatible
  • Flexible enough to permit use of single size with several different needle gauges; yet sufficiently rigid to minimize danger of wall collapse
  • Gas, chemical, and gamma sterilizable
  • Durometer hardness: Shore A, 80
On reel of 500 ft. (152.4m)
Meets U.S. Pharmacopeia Class VI criteria for medical plastics.