Please login to your online account to display your discounted pricing
Saint-Gobain™ Tygon™ E-1000 Non-DEHP Laboratory Tubing
$99.18 - $777.85
Specifications
Length (English) 50 ft.
Length (Metric) 15m
View More Specs
Products
Catalog Number Mfr. No. Diameter (Metric) Inner Pressure Thickness Price Quantity    
14-171-118
saint-gobain™
ADK00012
4.8mm 10 psi Pack of 50 Each for $207.58
14-171-117
saint-gobain™
ADK00007
3.2mm 16 psi Pack of 50 Each for $145.06
14-171-116
saint-gobain™
ADK00003
1.6mm 26 psi Pack of 50 Each for $99.18
14-171-121
saint-gobain™
ADK00036
12.7mm 6 psi Pack of 50 Each for $411.12
14-171-122
saint-gobain™
ADK00038
12.7mm 8 psi Pack of 50 Each for $777.85
14-171-120
saint-gobain™
ADK00029
9.5mm 11 psi Pack of 50 Each for $629.36
Description & Specifications
Specifications
Length (English) 50 ft.
Length (Metric) 15m