Saint-Gobain Norwell™ Mix-and-Match Bailer Components Complete Assemblies (1050cc capac.)
Ideal for dedicated or portable applications
Manufacturer:  Saint-Gobain D8653025
 View more versions of this product
Catalog No. 15-437-50F
Description
Description
Allow you to choose the right combination of components to adapt to changing sampling needs and provide a flexible, cost-effective system for monitoring groundwater contamination near landfills and other pollution sources. Precision-machined, interchangeable parts allow quick reassembly after cleaning.
  • Made of 100% virgin PTFE and FEP
  • Minimize the need for retesting
  • Chemically inert and free of fillers and plasticizers
  • Nonleaching surface resists corrosion and minimizes sample bias
Order Info
Components can be ordered individually, in various combinations, or as part of fully assembled bailer
1All assemblies come with either 1 or 3 ft. Bailer Body Extension Tube.
Specifications
Specifications
35.5 oz., 1050cc
3 ft.
Norwell™ Bailer Assembly
1050cc
Standard bailer top cap and bottom emptying check assembly
91.5cm
35.5 oz.
FEP and PTFE
Documents