Saint-Gobain™ Norprene™ Chemical Tubing
$382.09 - $392.82
Specifications
Material Norprene
Thickness 0.0625 in.
Thickness (English) Wall 0.0625 in.
View More Specs
Products ${productFamilyLength}
Catalog Number Mfr. No. Diameter (English) Inner Diameter (English) Outer Max. Pressure Price Quantity  
Catalog Number Mfr. No. Diameter (English) Inner Diameter (English) Outer Max. Pressure Price Quantity  
14-168-225 Saint-Gobain™
AD300007
0.13 in. 0.25 in. 33psi Pack for $382.09
14-168-226 Saint-Gobain™
AD300012
0.188 in. 0.3125 in. 29psi Pack for $392.82
Specifications Description & Specifications
Specifications
Norprene
0.0625 in.
0.0625 in.