Please login to your online account to display your discounted pricing

Chemware™ PTFE Griffin Beakers


Noncontaminating and inert to highly corrosive reagents such as hydrofluoric acid

Manufacturer: saint gobain perform plas  D1069033

 View more versions of this product

Catalog No. 02-586-1B

  • / Each
 

Description & Specifications

Specifications

Product Type Beaker
Material PTFE (Polytetrafluoroethylene)
Includes Spout
Capacity (Metric) 3mL
For Use With (Application) Ideal for digestion
Height (English) 0.5 in.
Height (Metric) 1.27cm
Temperature (English) Max. Operating 550°F
Temperature (Metric) Max. Operating 288°C