Please login to your online account to display your discounted pricing

Saint-Gobain™ Chemware™ PTFE Griffin Beakers


Noncontaminating and inert to highly corrosive reagents such as hydrofluoric acid

Manufacturer: saint-gobain™  D1069032

 View more versions of this product

Catalog No. 02-586-1A

  • / Each
 

Description & Specifications

Specifications

Product Type Beaker
Includes Spout
Material PTFE (Polytetrafluoroethylene)
Capacity (Metric) 1mL
Height (English) 0.5 in.
Height (Metric) 1.27cm
For Use With (Application) Ideal for digestion
Temperature (English) Max. Operating 550°F
Temperature (Metric) Max. Operating 288°C