Please login to your online account to display your discounted pricing

Saint-Gobain™ Chemware™ PTFE Griffin Beakers


Noncontaminating and inert to highly corrosive reagents such as hydrofluoric acid

Manufacturer: saint-gobain™  D1069037

 View more versions of this product

Catalog No. 02-586-1G

  • / Each
 

Description & Specifications

Specifications

Product Type Beaker
Material PTFE (Polytetrafluoroethylene)
Includes Spout
Capacity (Metric) 100mL
For Use With (Application) Ideal for digestion
Height (English) 2.57 in.
Temperature (English) Max. Operating 550°F
Height (Metric) 6.55cm
Temperature (Metric) Max. Operating 288°C