Please login to your online account to display your discounted pricing

Saint-Gobain™ Chemware™ PTFE Griffin Beakers


Noncontaminating and inert to highly corrosive reagents such as hydrofluoric acid

Manufacturer: saint-gobain™  D1069035

 View more versions of this product

Catalog No. 02-586-1E

  • / Each
 

Description & Specifications

Specifications

Product Type Beaker
Material PTFE (Polytetrafluoroethylene)
Includes Spout
Capacity (Metric) 30mL
For Use With (Application) Ideal for digestion
Height (English) 1.77 in.
Height (Metric) 4.5cm
Temperature (English) Max. Operating 550°F
Temperature (Metric) Max. Operating 288°C