Saint-Gobain™ Chemware™ PTFE Griffin Beakers
Noncontaminating and inert to highly corrosive reagents such as hydrofluoric acid
Manufacturer:  Saint-Gobain™ D1069034
 View more versions of this product
Catalog No. 02-586-1C
Specifications Description & Specifications
Specifications
Beaker
Spout
288°C
Ideal for digestion
0.99 in.
PTFE (Polytetrafluoroethylene)
5mL
550°F
2.53cm