Saint-Gobain™ Chemware™ FEP Gas Sampling Bags
5mL thick FEP film for low permeability and maximum sample retention
Manufacturer:  Saint-Gobain™ D107500610
 View more versions of this product
Catalog No. 10-923-6
Specifications Description & Specifications
Specifications
6 x 10 in. (15 x 25cm)
Septum valve
Gas Sampling Bag
0.127mm
40.57 oz.
15cm
25cm
For gas sample analysis, extracting air samples from exhaust systems
6 in.
5 mil
10 in.
1.2L
FEP (Fluorinated Ethylene Propylene)