Please login to your online account to display your discounted pricing
Saint-Gobain™ Chemware™ Bendable PTFE Stirring Rods
Contain fully enclosed steel insert for stiffness, yet can be bent to various shapes for use as powered stirrers
$409.26 - $415.29
Products
Catalog Number Mfr. No. Price Quantity    
14-518-10F
saint-gobain™
D1069091
Each for $415.29
14-518-10E
saint-gobain™
D1069089
Each for $409.26
Description & Specifications
  • Heat resistant to 288°C (550°F)
  • Completely inert to highly corrosive reagents
  • Anti-stick properties facilitate cleaning
  • Unbreakable