Saint-Gobain™ Bio-Simplex™ Polypropylene Carboys
Manufacturer:  Saint-Gobain™ DCB20120
 View more versions of this product
Catalog No. 02-611-205
Specifications Description & Specifications
Specifications
±5%
Liquid handling and solution storage for general laboratory, process development and bioproduction applications
676 oz.
Bottom handles for full hand grip and closed cap
Carboy
Polypropylene
21 CFR 177, USP Class VI
676.28 oz., 20L
Wide
20L
676.28 oz.
20L
Yes