Saint-Gobain™ Bio-Simplex™ Polypropylene Carboys
Manufacturer:  Saint-Gobain™ DCB20120S
 View more versions of this product
Catalog No. 02-611-206
Specifications Description & Specifications
Specifications
676 oz.
Liquid handling and solution storage for general laboratory, process development and bioproduction applications
676.28 oz., 20L
20L
±5%
21 CFR 177, USP Class VI
Yes
Bottom handles for full hand grip, closed cap and spigot
20L
Wide
Carboy
676.28 oz.
Polypropylene