Saint-Gobain™ Bio-Simplex™ Polypropylene Carboys
$287.67 - $1,266.21
Specifications
Accuracy ±5%
Certifications/Compliance 21 CFR 177, USP Class VI
Autoclavable Yes
Material Polypropylene
Product Type Carboy
View More Specs
Products ${productFamilyLength}
Catalog Number Mfr. No. Capacity With Price Quantity  
Catalog Number Mfr. No. Capacity With Price Quantity  
02-611-201 Saint-Gobain™
DCB0580S
169.07 oz., 5L Bottom handles for full hand grip, closed cap and spigot Each for $367.58
02-611-202 Saint-Gobain™
DCB1080
338.14 oz., 10L Bottom handles for full hand grip and closed cap Each for $380.73
02-611-200 Saint-Gobain™
DCB0580
169.07 oz., 5L Bottom handles for full hand grip and closed cap Each for $287.67
02-611-204 Saint-Gobain™
DCB10S3
338.14 oz., 10L Each for $639.48
02-611-203 Saint-Gobain™
DCB1080S
338.14 oz., 10L Bottom handles for full hand grip, closed cap and spigot Each for $486.06
02-611-205 Saint-Gobain™
DCB20120
676.28 oz., 20L Bottom handles for full hand grip and closed cap Each for $442.33
02-611-207 Saint-Gobain™
DCB20S3
676.28 oz., 20L Each for $1,149.33
02-611-209 Saint-Gobain™
DCB40120S
1352.56 oz., 40L Bottom handles for full hand grip, closed cap and spigot Each for $881.39
02-611-206 Saint-Gobain™
DCB20120S
676.28 oz., 20L Bottom handles for full hand grip, closed cap and spigot Each for $661.09
02-611-208 Saint-Gobain™
DCB40120
1352.56 oz., 40L Bottom handles for full hand grip and closed cap Each for $659.67
02-611-210 Saint-Gobain™
DCB40S3
1352.56 oz., 40L Each for $1,266.21
Specifications Description & Specifications
Specifications
±5%
Yes
Carboy
Wide
21 CFR 177, USP Class VI
Polypropylene
Liquid handling and solution storage for general laboratory, process development and bioproduction applications