Saint-Gobain ALUNDUM™ Filtering Crucibles
For melting metals and vaporizing liquids
Manufacturer:  Saint-Gobain 5205AL889
 View more versions of this product
Catalog No. 08-230B
Description
Description
  • Not affected by acids or alkalis
  • Porous for filtering with suction and holding finest precipitates
  • Dry quickly to constant weight
  • Available in medium, made from AN889 ALUNDUM; coarse, made from AN485 mix
Specifications
Specifications
Medium Grain Alundum™
35mL
43mm
Medium
For melting metals and vaporizing liquids
41mm
Alundum