Saint-Gobain ALUNDUM™ Extraction Thimbles
Wash, ignite, and use repeatedly
Manufacturer:  Norton Company AN4856406
 View more versions of this product
Catalog No. 09-661E
Specifications Description & Specifications
Specifications
Extraction Thimble
20μm
Alundum™
10cm
3.93 in.
Used for all kinds of extractions except strong alkalis
1.33 in.
Round
3.4cm
Coarse