Saint-Gobain ALUNDUM™ Extraction Thimbles
Wash, ignite, and use repeatedly
$242.52 - $2,036.00
Specifications
Material Alundum™
For Use With (Application) Used for all kinds of extractions except strong alkalis
Product Type Extraction Thimble
View More Specs
Products 3
Catalog Number Mfr. No. Bottom Shape Height (English) Pore Size Diameter (Metric) Outer Price Quantity  
Catalog Number Mfr. No. Bottom Shape Height (English) Pore Size Diameter (Metric) Outer Price Quantity  
09-660E
View Documents
Saint-Gobain
6945AL889
Round 5 in. 5μm 4.5cm
Case of 6 Each for $2,036.00
09-661E
View Documents
Saint-Gobain
AN4856406
Round 3.93 in. 20μm 3.4cm
Each for $242.52
Case of 6 Each for $1,455.12
09-661F
View Documents
Saint-Gobain
6945AL485
Round 5 in. 20μm 4.5cm
Each for $409.66
Case of 6 Each for $2,137.38
Description
Description
  • Used for all kinds of extractions except strong alkalis
  • ALUNDUM AN889 thimbles (Series 09-660) have medium porosity, retain particles approximately 5μm and larger
  • ALUNDUM AN485 thimbles (Series 09-661) have coarse porosity for faster extraction, less retentivity; retain particles approximately 20μm and larger
  • 34 and 45mm diameter thimbles in each series have round bottoms; all other sizes have flat bottoms
†With round bottoms.
Specifications
Specifications
Alundum™
Used for all kinds of extractions except strong alkalis
Extraction Thimble
SDS