Please login to your online account to display your discounted pricing

Safranine, Aqueous-Alcoholic, Counterstain for Gram Staining, Ricca Chemical

$295.03 - $295.03

Chemical Identifiers

Chemical Name or Material Safranine
Name Note Counterstain for Gram Staining
CAS 7732-18-5
CAS Min % 90.0
CAS Max % 94.0
CAS 64-17-5
CAS Min % 7.24
CAS Max % 7.53
CAS 477-73-6
CAS Min % 0.32
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

6680-1

 
ricca chemical company
RI66801
Poly natural 4L Each for $295.03
Description & Specifications

Specifications

Physical Form Liquid
Color Clear dark red
Quantity 4L
Solvent or Matrix Aqueous-Alcoholic
Name Note Counterstain for Gram Staining
CAS 477-73-6
CAS 7732-18-5
CAS 7732-18-5 (Water, Deionized)
CAS 64-17-5
CAS Min % 90.0
CAS Max % 94.0
CAS Min % 7.24
CAS Max % 7.53
CAS Max % 0.33
CAS Min % 0.32
Packaging Poly natural
Chemical Name or Material Safranine