Please login to your online account to display your discounted pricing

Safranin O, Reagent Grade, LabChem™

$144.40 - $144.40

Chemical Identifiers

Chemical Name or Material Safranin O
CAS 477-73-6
CAS Max % 100
Purity Grade Reagent Grade
Linear Formula C20H19ClN4
Formula Weight 350.85
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

LC223408

 
lab chem
LC223408
Amber Glass 25g Each for $144.40
Description & Specifications

Specifications

Formula Weight 350.85
Chemical Name or Material Safranin O
Purity Grade Reagent Grade
Physical Form Powder
Identification Passes Test
Linear Formula C20H19ClN4
CAS 477-73-6
Packaging Amber Glass
Health Hazard 1 Danger
Solubility Soluble in water
CAS Max % 100
Health Hazard 2 GHS H Statement
Causes skin irritation and serious eye damage.
Storage Information Room Temperature
Health Hazard 3 GHS P Statement
Wear protective gloves, eye protection.
Wash exposed skin thoroughly after handling.
If on skin: Wash with plenty of soap and water.
If skin irritation occurs: Get medical attention.
Take off contaminated clothing and wash before reuse.
If in eyes: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
Immediately call a poison center/doctor.
Decomposition Information Nitrogen oxides; Carbon monoxide; Carbon dioxide
Color Dark green
Quantity 25g
Safety and Handling

Danger

GHS P Statement
Wear protective gloves, eye protection.
Wash exposed skin thoroughly after handling.
If on skin: Wash with plenty of soap and water.
If skin irritation occurs: Get medical attention.
Take off contaminated clothing and wash before reuse.
If in eyes: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
Immediately call a poison center/doctor.

GHS H Statement
Causes skin irritation and serious eye damage.