Please login to your online account to display your discounted pricing

Safranin, 1% (w/v) Alcoholic, Certified, LabChem™

$56.91 - $56.91

Chemical Identifiers

Chemical Name or Material Safranin
CAS 64-17-5
CAS Max % 89.1
CAS 67-63-0
CAS Max % 4.95
CAS 67-56-1
CAS Max % 4.95
CAS 477-73-6
CAS Max % 1
Name Note 1% (w/v) Alcoholic
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

LC223601

 
lab chem
LC223601
Poly Bottle 500mL Each for $56.91
Description & Specifications

Specifications

Purity Grade Certified Grade
Formula Weight 350.85
CAS 477-73-6
Flash Point 13°C
CAS Max % 4.95
Color Dark blood-red
CAS 67-63-0
Chemical Name or Material Safranin
CAS 64-17-5
Boiling Point 78°C
Name Note 1% (w/v) Alcoholic
UN Number UN1170
Packaging Poly Bottle
Health Hazard 1 Danger
Health Hazard 2 GHS H Statement
Highly flammable liquid and vapor.
Causes serious eye irritation.
May cause cancer.
Suspected of damaging fertility or the unborn child.
Causes damage to organs (central nervous system, optic nerve, liver, kidneys).
Physical Form Liquid
Solubility Soluble in water
CAS 67-56-1
CAS Max % 1
Linear Formula C20H19ClN4
Identification Passes Test
Storage Information Room Temperature
CAS Max % 4.95
CAS Max % 89.1
Health Hazard 3 GHS P Statement
Obtain special instructions before use.
Do not handle until all safety precautions have been read and understood.
Keep away from heat, sparks, open flames, hot surfaces.
No smoking.
Keep container tightly closed.
Ground/bond container and receiving equipment.
Use explosion-proof electrical, ventilating, lighting equipment.
Use only non-sparking tools.
Take precautionary measures against static discharge.
Do not breathe mist, vapors, spray.
Do not eat, drink or smoke when using this product.
Wear eye protection, protective gloves.
Wash exposed skin thoroughly after handling.
If exposed or concerned: Get medical advice/attention.
If on skin (or hair): Remove/Take off immediately all contaminated clothing.
Rinse skin with water/shower.
If in eyes: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
If eye irritation persists: Get medical advice/attention.
In case of fire: Use alcohol resistant foam, carbon dioxide (CO2), extinguishing powder to extinguish.
Store locked up in a cool, well-ventilated place.
Dispose of contents/container to comply with local, state and federal regulations.
Melting Point −115°C
Quantity 500mL
Decomposition Information Carbon monoxide; Carbon dioxide; Nitrogen oxides
Safety and Handling

Danger

GHS P Statement
Obtain special instructions before use.
Do not handle until all safety precautions have been read and understood.
Keep away from heat, sparks, open flames, hot surfaces.
No smoking.
Keep container tightly closed.
Ground/bond container and receiving equipment.
Use explosion-proof electrical, ventilating, lighting equipment.
Use only non-sparking tools.
Take precautionary measures against static discharge.
Do not breathe mist, vapors, spray.
Do not eat, drink or smoke when using this product.
Wear eye protection, protective gloves.
Wash exposed skin thoroughly after handling.
If exposed or concerned: Get medical advice/attention.
If on skin (or hair): Remove/Take off immediately all contaminated clothing.
Rinse skin with water/shower.
If in eyes: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
If eye irritation persists: Get medical advice/attention.
In case of fire: Use alcohol resistant foam, carbon dioxide (CO2), extinguishing powder to extinguish.
Store locked up in a cool, well-ventilated place.
Dispose of contents/container to comply with local, state and federal regulations.

GHS H Statement
Highly flammable liquid and vapor.
Causes serious eye irritation.
May cause cancer.
Suspected of damaging fertility or the unborn child.
Causes damage to organs (central nervous system, optic nerve, liver, kidneys).