Please login to your online account to display your discounted pricing

Saclofen, Tocris


Selective GABAB antagonist

Quantity 10mg

Manufacturer: r & d systems  RDS024610

 View more versions of this product

Catalog No. 024610

  • / Each
 

Description & Specifications

Chemical Identifiers

Quantity 10mg
Formula Weight 249.72
Chemical Name or Material Saclofen
CAS 125464-42-8
Assay Percent Range >99%
Molecular Formula C9H12ClNO3S
Synonym (RS)-3-Amino-2-(4-chlorophenyl)propylsulfonic acid
Recommended Storage Store at Room Temperature

Specifications

Chemical Name or Material Saclofen
Assay Percent Range >99%
Molecular Formula C9H12ClNO3S
Formula Weight 249.72
Synonym (RS)-3-Amino-2-(4-chlorophenyl)propylsulfonic acid
Accessories

Store at Room Temperature