missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Saccomanno Collection Fluid, Fixative and Preservative for Sputum Samples, Ricca Chemical

Chemical Identifiers

CAS 7732-18-5,64-17-5,25322-68-3,1934-21-0,3844-45-9
CAS Min % 53.0,42.28,2.3,5.0E-4,4.0E-4
CAS Max % 56.0,44.01,2.4,5.0E-4,4.0E-4
Physical Form Liquid
Chemical Name or Material Saccomanno Collection Fluid, Fixative and Preservative for Sputum Samples
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Packaging Quantity Price Quantity  
Catalog Number Mfr. No. Packaging Quantity Price Quantity  
6655-1 Ricca Chemical Company
RI66551
Poly natural 4L N/A N/A  
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
6655-5 SDS Ricca Chemical Company
RI66555
Cubitainer 20L N/A N/A  
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Specifications Description & Specifications

Chemical Identifiers

7732-18-5,64-17-5,25322-68-3,1934-21-0,3844-45-9
56.0,44.01,2.4,5.0E-4,4.0E-4
Saccomanno Collection Fluid, Fixative and Preservative for Sputum Samples
53.0,42.28,2.3,5.0E-4,4.0E-4
Liquid

Specifications

4L
53.0,42.28,2.3,5.0E-4,4.0E-4
Liquid
Clear green
7732-18-5,64-17-5,25322-68-3,1934-21-0,3844-45-9
56.0,44.01,2.4,5.0E-4,4.0E-4
Poly natural
Saccomanno Collection Fluid, Fixative and Preservative for Sputum Samples