missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Saccharin, sodium salt hydrate, 99+%, ACROS Organics™

Manufacturer:  Fisher Scientific Acros 223372500

 View more versions of this product

Catalog No. AC223372500


 

Specifications Description & Specifications

Specifications

Saccharin, sodium salt hydrate
82385-42-0
3ppm max.
20ppm max.
99.0 to 101.0% (on anhydrous substance)
C7H4NNaO3S·xH2O
205.16
Saccharin soluble
Passes Test
15% max.
250g
99+%
toluenesulfonamides (GC): < 25ppm
Passes Test
Crystals or Crystalline Powder
99+%
Authentic
MFCD00149605
Plastic bottle
30ppm max.
Colorless to white

Chemical Identifiers

82385-42-0
C7H4NNaO3S·xH2O
MFCD00149605
Saccharin, sodium salt hydrate, 99+%
99+%
205.16
Saccharin soluble
99.0 to 101.0% (on anhydrous substance)