Please login to your online account to display your discounted pricing

Saccharin Sodium (Powder/USP/FCC), Fisher Chemical


Quantity 500g
Packaging Poly Bottle

Manufacturer: fisher scientific company  FLS394500

 View more versions of this product

Catalog No. S394-500

  • / Each
 

Description & Specifications

Chemical Identifiers

Purity Grade FCC/USP
Formula Weight 205.17
Quantity 500g
Assay 98.0 to 101.0% (USP)
Physical Form Solid
Molecular Formula C7H4NO3S · Na · 2 H2O
Color White
Water 15% Max. (FCC/USP)
Selenium (Se) 0.003% max. (FCC)
Residual Solvents Meets Requirements (USP)
Lead (Pb) 2mg/kg max. (FCC)
o_Toluenesulfonamide 10ppm max. (USP)
Benzoate and Salicylate Pass Test (FCC)
Carbonizable Substances Pass Test (FCC)
Clarity Pass Test (USP)
Packaging Poly Bottle
Identification Pass Test (FCC)
ChemAlert Storage Symbol Gray
MDL Number MFCD00013092
Toluene Sulfonamides 0.0025% max. (FCC)
p_Toluenesulfonamide 10ppm max. (USP)
Heavy Metals (as Pb) 0.001% max. (USP)
Acidity Alkalinity Pass Test (USP)
Alkalinity Pass Test (FCC)
CAS 128-44-9
Chemical Name or Material Saccharin Sodium

Specifications

Chemical Name or Material Saccharin Sodium
CAS 128-44-9
Purity Grade FCC/USP
Physical Form Solid
Assay 98.0 to 101.0% (USP)
Molecular Formula C7H4NO3S · Na · 2 H2O
Formula Weight 205.17
MDL Number MFCD00013092
Accessories
 

SDS & Safety Documents