Please login to your online account to display your discounted pricing

Saccharin Sodium (Powder/USP/FCC), Fisher Chemical


Quantity 12kg
Packaging Poly Pail

Manufacturer: fisher scientific company  FLS39412

 View more versions of this product

Catalog No. S394-12

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Purity Grade FCC/USP
Formula Weight 205.17
Quantity 12kg
Physical Form Solid
Assay 98.0 to 101.0% (FCC)
Color White
Molecular Formula C7H4NO3S · Na · 2 H2O
Water 15% Max. (FCC/USP)
Selenium (Se) 0.003% max. (FCC)
Heavy Metals (as Pb) 0.001% max. (USP)
Carbonizable Substances Pass Test (FCC)
Lead (Pb) 2mg/kg max. (FCC)
Benzoate and Salicylate Pass Test (FCC)
MDL Number MFCD00013092
ChemAlert Storage Symbol Gray
o_Toluenesulfonamide 10ppm max. (USP)
Alkalinity Pass Test (FCC)
Identification Pass Test (FCC)
Packaging Poly Pail
Acidity Alkalinity Pass Test (USP)
Clarity Pass Test (USP)
Toluene Sulfonamides 0.0025% max. (FCC)
p_Toluenesulfonamide 10ppm max. (USP)
Residual Solvents Meets Requirements (USP)
CAS 128-44-9
Chemical Name or Material Saccharin Sodium

Specifications

Chemical Name or Material Saccharin Sodium
CAS 128-44-9
Purity Grade FCC/USP
Physical Form Solid
Assay 98.0 to 101.0% (FCC)
Molecular Formula C7H4NO3S · Na · 2 H2O
Formula Weight 205.17
MDL Number MFCD00013092
Accessories
 

SDS & Safety Documents