missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Saccharin, 98+%, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals 149001000
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Saccharin | |
226.0°C to 230.0°C | |
1% (105°C, 2 hrs) max. | |
98+% | |
C7H5NO3S | |
27,168 | |
saccharin, saccharine, o-sulfobenzimide, o-benzoic sulfimide, saccharimide, benzosulfimide, o-benzosulfimide, benzoic sulfimide, benzosulphimide, saccharinose | |
CVHZOJJKTDOEJC-UHFFFAOYSA-N | |
1,1-dioxo-1,2-benzothiazol-3-one | |
5143 | |
183.18 | |
Crystalline Powder or Crystals |
81-07-2 | |
White | |
Authentic | |
Plastic bottle | |
100 g | |
15,8445 | |
Solubility in water: 3.3g/L (20°C). Other solubilities: 1g/31mL alcohol - 1g/12mL acetone, 1g/50mL glycerol, soluble in amyl acetate and ethyl acetate, slightly soluble in chloroform and ether, soluble in dil. solution of ammonia | |
C1=CC=C2C(=C1)C(=O)NS2(=O)=O | |
183.18 | |
CHEBI:32111 | |
98+% |
Chemical Identifiers
81-07-2 | |
183.18 | |
saccharin, saccharine, o-sulfobenzimide, o-benzoic sulfimide, saccharimide, benzosulfimide, o-benzosulfimide, benzoic sulfimide, benzosulphimide, saccharinose | |
CHEBI:32111 | |
C1=CC=C2C(=C1)C(=O)NS2(=O)=O |
C7H5NO3S | |
CVHZOJJKTDOEJC-UHFFFAOYSA-N | |
5143 | |
1,1-dioxo-1,2-benzothiazol-3-one |
Safety and Handling
EINECSNumber : 201-321-
RUO – Research Use Only