Please login to your online account to display your discounted pricing

Thermo Scientific™ Sabouraud Dextrose Agar (Dehydrated)


Identify dermatophytes and other fungi and yeasts with the acidic Thermo Scientific™ Oxoid™ Sabouraud Dextrose Agar.

Manufacturer: thermo scientific™  CM0041B

 View more versions of this product

Catalog No. OXCM0041B

  • / Each
 

Description & Specifications

Specifications

Format Powder
Type Media
Quantity 500g
Description Sabouraud Dextrose Agar
Storage Requirements 10-30°C
Shelf Life 5 years
Item Description Sabouraud Dextrose Agar; 500g
Packaging Each
Yield For 7.7L medium

Identify dermatophytes and other fungi and yeasts in clinical specimens and environmental samples with the acidic Thermo Scientific™ Oxoid™ Sabouraud Dextrose Agar. The low pH supports the growth of fungi while inhibiting bacterial growth in mixed samples. Fungi maintain their typical cultural appearance and, thus, may be readily identified according to standard macroscopic characters.