Please login to your online account to display your discounted pricing

SA 57, Tocris™

Potent FAAH inhibitor

$201.36 - $844.73

Chemical Identifiers

Chemical Name or Material SA 57
Assay Percent Range >99%
Molecular Formula C17H23ClN2O3
Formula Weight 338.83
Synonym 4-[2-(4-Chlorophenyl)ethyl]-1-piperidinecarboxylic acid 2-(methylamino)-2-oxoethyl ester
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

475210

 
r & d systems
RDS475210
10mg Each for $201.36

475250

 
r & d systems
RDS475250
50mg Each for $844.73
Description & Specifications

Specifications

Formula Weight 338.83
Quantity 10mg
Chemical Name or Material SA 57
Assay Percent Range >99%
Molecular Formula C17H23ClN2O3
CAS 1346169-63-8
Synonym 4-[2-(4-Chlorophenyl)ethyl]-1-piperidinecarboxylic acid 2-(methylamino)-2-oxoethyl ester
Recommended Storage Store at +4°C
Safety and Handling

Store at +4°C