Please login to your online account to display your discounted pricing

SA 47, Tocris™

Selective FAAH inhibitor

$188.60 - $794.96

Chemical Identifiers

Chemical Name or Material SA 47
Assay Percent Range >99%
Molecular Formula C17H26N4O3
Formula Weight 334.41
Synonym N-[2-[1-(6-Methyl-2-pyridinyl)-4-piperidinyl]ethyl]carbamic acid 2-(methylamino)-2-oxoethyl ester
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

478510

 
r & d systems
RDS478510
10mg Each for $188.60

478550

 
r & d systems
RDS478550
50mg Each for $794.96
Description & Specifications

Specifications

Formula Weight 334.41
Quantity 10mg
Chemical Name or Material SA 47
Assay Percent Range >99%
Molecular Formula C17H26N4O3
CAS 792236-07-8
Synonym N-[2-[1-(6-Methyl-2-pyridinyl)-4-piperidinyl]ethyl]carbamic acid 2-(methylamino)-2-oxoethyl ester
Recommended Storage Store at -20°C
Safety and Handling

Store at -20°C