Please login to your online account to display your discounted pricing

Tocris™ S26948

Selective PPARgamma agonist; antidiabetic agent

$252.39 - $1,050.42

Specifications

Product Type S26948
Inhibitors S26948
Molecular Weight 519.57
Purity >99
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Quantity Price Quantity    

369010R

 
tocris™
RDS369010
10mg Each for $252.39

369050R

 
tocris™
RDS369050
50mg Each for $1,050.42
Description & Specifications

Specifications

Product Type S26948
Inhibitors S26948
Molecular Weight 519.57
Purity >99

Selective PPARγ agonist (EC50 = 8.83nM). Drives normal adipocyte differentiation and exhibits a lipid-lowering effect; reduces atherosclerosis in E2-KI mice. Displays antidiabetic properties and improves hepatic insulin sensitivity in intralipid-infusion (IL) rats.

  • Chemical Name: [[4-[2-(6-Benzoyl-2-oxo-3(2H)-benzothiazolyl)ethoxy]phenyl]methyl]-1,3-propanedioic acid dimethyl ester
  • Formula: C28H25NO7S
  • CAS Number: 353280-43-0
  • Solubility: Soluble to 100mM in DMSO
  • Purity: >99