Learn More
S100A8 Monoclonal Antibody (CF-145), eFluor™ 660, eBioscience™, Invitrogen™
Mouse Monoclonal Antibody
$179.00 - $179.00
Specifications
Antigen | S100A8 |
---|---|
Clone | CF-145 |
Applications | Flow Cytometry |
Classification | Monoclonal |
Conjugate | eFluor 660 |
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
501124521
|
Invitrogen
50974541 |
25 Tests |
N/A
|
|
|||||
Description
The protein encoded by this gene is a member of the S100 family of proteins containing 2 EF-hand calcium-binding motifs. S100 proteins are localized in the cytoplasm and/or nucleus of a wide range of cells, and involved in the regulation of a number of cellular processes such as cell cycle progression and differentiation. S100 genes include at least 13 members which are located as a cluster on chromosome 1q21. This protein may function in the inhibition of casein kinase and as a cytokine. Altered expression of this protein is associated with the disease cystic fibrosis.
S100A8 Monoclonal antibody specifically detects S100A8 in Human samples. It is validated for Flow Cytometry.Specifications
S100A8 | |
Flow Cytometry | |
eFluor 660 | |
Mouse | |
Human | |
6279 | |
IgG2b κ | |
Affinity chromatography | |
S100A8 |
CF-145 | |
Monoclonal | |
Liquid | |
RUO | |
P05109 | |
S100A8 | |
Primary | |
Antibody |
For Research Use Only.