missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

S100A8 Mouse anti-Human, Clone: CF-145, eBioscience™

Mouse Monoclonal Antibody

$131.95 - $339.30

Specifications

Antigen S100A8
Clone CF-145
Host Species Murine
Gene Alias Calgranulin A, Calprotectin
Species Reactivity Human
View More Specs

Disclaimers:

For Research Use Only.



Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
501122813 Life Technologies
LS14974580
25μg Each for $131.95
501122814 Life Technologies
LS14974582
100μg Each for $339.30
Specifications

Description

The monoclonal antibody CF-145 recognizes human S100A8. S100A8, also known as migration inhibitory factor-related protein 8 (MRP-8) or Calgranulin-A, is a calcium binding protein and member of the S100 family of proteins. S100A8 is expressed by neutrophils, monocytes, macrophages, granulocytes, and epithelial cells during dermatoses, such as psoriasis. S100A8 forms a heterodimer with S100A9, which together is known as calprotectin. S100A8 has antimicrobial and pro-apoptotic properties and is a pro-inflammatory mediator in acute and chronic inflammation. Levels of S100A8 are elevated in patients with various inflammatory disorders such as cystic fibrosis, rheumatoid arthritis, and chronic bronchitis. Upregulation of S100A8 is also observed in many tumors, including skin, breast, colon, pancreatic, and ovarian cancers.
Description & Specifications

Specifications

S100A8
Murine
Human
RUO
Purified
Primary
aqueous buffer, 0.09% sodium azide, may contain carrier protein/stabilizer
CF-145
Calgranulin A, Calprotectin
Immunohistochemistry (Formalin/Paraffin),Western Blot
Unlabeled
Store at 2-8°C.
Monoclonal
0.5mg/mL