missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Thermo Scientific™ Lab Vision™ S100 A6 Ab-9, Rabbit Polyclonal Antibody

Provide accurate, reproducible results with the Thermo Scientific™ S100 A6 Ab-9, Rabbit Polyclonal Antibody.

$255.18 - $1,402.47

Specifications

Antigen S100 A6 Ab-9
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Conjugate Unlabeled
Cross Reactivity Bovine,Canine,Equine,Feline,Human,Mouse,Bovine,Canine,Equine,Feline,Human,Mouse,Bovine,Canine,Equine,Feline,Human,Mouse,Bovine,Canine,Equine,Feline,Human,Mouse,Bovine,Canine,Equine,Feline,Human,Mouse,Bovine,Canine,Equine,Feline,Human,Mouse
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
RB1805A Thermo Scientific™ Lab Vision™
RB1805A
1mL Each for $1,402.47
RB1805A0 Thermo Scientific™ Lab Vision™
RB1805A0
100μL Each for $255.18
RB1805A1 Thermo Scientific™ Lab Vision™
RB1805A1
500μL Each for $979.98
RB1805R7 Thermo Scientific™ Lab Vision™
RB1805R7
7mL Each for $683.55
Specifications

Description

Provide accurate, reproducible results with the Thermo Scientific™ S100 A6 Ab-9, Rabbit Polyclonal Antibody.

S100 belongs to the family of calcium binding proteins such as calmodulin and troponin C. S100A is composed of an α and β chain whereas S100B is composed of two β chains. S100 protein is also expressed in the antigen presenting cells such as the Langerhans cells in skin and interdigitating reticulum cells in the paracortex of lymph nodes.

Host Species: Rabbit

Isotype: IgG

Species Reactivity: Human, Mouse, Rat, Cow, Horse, Cat and Dog. Others not known.

Epitope: Not determined

Immunogen: Purified recombinant human S100 A6 monomer expressed in E. coli

Molecular Weight: 10-12kDa

Positive Control: Tonsil

Cellular Localization: Cytoplasmic

Recommended for:

  • Immunohistochemistry (Formalin/paraffin)

Note:

USA: RUO; Int'l: RUO

Description & Specifications

Specifications

S100 A6 Ab-9
Polyclonal
Bovine,Canine,Equine,Feline,Human,Mouse,Bovine,Canine,Equine,Feline,Human,Mouse,Bovine,Canine,Equine,Feline,Human,Mouse,Bovine,Canine,Equine,Feline,Human,Mouse,Bovine,Canine,Equine,Feline,Human,Mouse,Bovine,Canine,Equine,Feline,Human,Mouse
RUO
Primary
IgG
Immunohistochemistry (Paraffin)
Unlabeled
Rabbit
Neuroscience
Purified recombinant human S100 A6 monomer expressed in E. coli