Please login to your online account to display your discounted pricing

Thermo Scientific™ Lab Vision™ S100 A4, Rabbit Polyclonal Antibody

Provide accurate, reproducible results with the Thermo Scientific™ S100 A4, Rabbit Polyclonal Antibody.

$277.63 - $1,291.12

Specifications

Antigen S100 A4
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Conjugate Unlabeled
Cross Reactivity Human
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

RB9411P

 
thermo scientific™ lab vision™
RB9411P
1mL Each for $1,291.12

RB9411P0

 
thermo scientific™ lab vision™
RB9411P0
100µL Each for $277.63

RB9411P1

 
thermo scientific™ lab vision™
RB9411P1
500µL Each for $804.39

RB9411R7

 
thermo scientific™ lab vision™
RB9411R7
7mL Each for $578.06
Description & Specifications

Specifications

Antigen S100 A4
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Conjugate Unlabeled
Cross Reactivity Human
Host Species Rabbit
Regulatory Status RUO
Research Discipline Neuroscience
Type Primary
Immunogen Recombinant protein encoding full length human S100A4

Provide accurate, reproducible results with the Thermo Scientific™ S100 A4, Rabbit Polyclonal Antibody.

S100 belongs to the family of calcium binding proteins such as calmodulin and troponin C. S100A is composed of an alpha and beta chain whereas S100B is composed of two beta chains. S100 protein is also expressed in the antigen presenting cells such as the Langerhans cells in skin and interdigitating reticulum cells in the paracortex of lymph nodes.

Host Species: Rabbit

Species Reactivity: Human. Others not tested.

Immunogen: Recombinant protein encoding full-length human S100 A4

Molecular Weight: 10-12kDa

Positive Control: Human melanoma

Cellular Localization: Cytoplasmic

Recommended for:

  • Immunohistochemistry (Formalin/paraffin)

Note:

USA: RUO; Int'l: RUO