Please login to your online account to display your discounted pricing

(S)-WAY 100135 dihydrochloride, Tocris


Potent, selective 5-HT1A antagonist

Quantity 10mg

Manufacturer: r & d systems  125310

 View more versions of this product

Catalog No. 125310

  • / Each
 

Description & Specifications

Chemical Identifiers

Formula Weight 468.47
Quantity 10mg
Assay Percent Range >99%
Chemical Name or Material (S)-WAY 100135 dihydrochloride
Molecular Formula C24H33N3O2·2HCl
CAS 149007-54-5
Synonym (S)-N-tert-Butyl-3-(4-(2-methoxyphenyl)-piperazin-1-yl)-2-phenylpropanamide dihydrochloride
Recommended Storage Desiccate at +4°C

Specifications

Chemical Name or Material (S)-WAY 100135 dihydrochloride
Assay Percent Range >99%
Molecular Formula C24H33N3O2·2HCl
Formula Weight 468.47
Synonym (S)-N-tert-Butyl-3-(4-(2-methoxyphenyl)-piperazin-1-yl)-2-phenylpropanamide dihydrochloride
Accessories

Desiccate at +4°C