Please login to your online account to display your discounted pricing

(S)-WAY 100135 dihydrochloride, Tocris

Potent, selective 5-HT1A antagonist

$201.36 - $844.73

Chemical Identifiers

Chemical Name or Material (S)-WAY 100135 dihydrochloride
Assay Percent Range >99%
Molecular Formula C24H33N3O2·2HCl
Formula Weight 468.47
Synonym (S)-N-tert-Butyl-3-(4-(2-methoxyphenyl)-piperazin-1-yl)-2-phenylpropanamide dihydrochloride
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

125310

 
r & d systems
125310
10mg Each for $201.36

125350

 
r & d systems
125350
50mg Each for $844.73
Description & Specifications

Specifications

Formula Weight 468.47
Quantity 10mg
Assay Percent Range >99%
Chemical Name or Material (S)-WAY 100135 dihydrochloride
Synonym (S)-N-tert-Butyl-3-(4-(2-methoxyphenyl)-piperazin-1-yl)-2-phenylpropanamide dihydrochloride
Recommended Storage Desiccate at +4°C
CAS 149007-54-5
Molecular Formula C24H33N3O2·2HCl
Safety and Handling

Desiccate at +4°C