Please login to your online account to display your discounted pricing

Alfa Aesar™ (+)-S-Trityl-L-cysteine, 97%

$28.32 - $65.09

Chemical Identifiers

Chemical Name or Material (+)-S-Trityl-L-cysteine
CAS 2799-07-7
Molecular Formula C22H21NO2S
Formula Weight 363.48
MDL Number MFCD00002611
Beilstein 2339626
Synonym H-Cys(Trt)-OH; S-Triphenylmethyl-L-cysteine
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAL1438403

  SDS
alfa aesar™
L1438403
1g Each for $28.32

AAL1438406

  SDS
alfa aesar™
L1438406
5g Each for $65.09
Description & Specifications

Specifications

Formula Weight 363.48
Chemical Name or Material (+)-S-Trityl-L-cysteine
CAS 2799-07-7
Health Hazard 1 Warning
Assay 97%
Molecular Formula C22H21NO2S
MDL Number MFCD00002611
Beilstein 2339626
Synonym H-Cys(Trt)-OH; S-Triphenylmethyl-L-cysteine
Health Hazard 2 GHS H Statement
H302
Harmful if swallowed.
Melting Point ≈185°C
Quantity 1g
Health Hazard 3 GHS P Statement
P264-P270-P301+P312-P330-P501a
Wash thoroughly after handling.
Do not eat, drink or smoke when using this product.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell. Rinse mouth.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Optical Rotation +84° (c=1 in Acetic Acid)
RTECS Number AY7710000
Safety and Handling

Warning

GHS P Statement
P264-P270-P301+P312-P330-P501a
Wash thoroughly after handling.
Do not eat, drink or smoke when using this product.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell. Rinse mouth.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H302
Harmful if swallowed.